Learn More
Molecular Devices SpectraMax™ ABS Single‐Mode Microplate Reader with SoftMax Pro Software
Shop All Molecular Devices ProductsDescription
- Designed for absorbance detection workflows in microplate-based assays
- Supports UV-Vis absorbance measurements for life science applications
- Enables flexible assay setup with configurable measurement parameters
- Supports microplate-based assay workflows across multiple applications
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Supports routine absorbance-based assays with consistent and reproducible performance
- Designed for use in laboratory environments requiring reliable microplate reading
Specifications
Specifications
| Detector Type | Photodiode |
| Interface | PC Control, Internal Software |
| Light Source | Xenon Flash Lamp |
| Measurement Technology | Absorbance (Top) |
| Plate Type | 96 and 384-well Plates |
| Reading Options | Endpoint |
| Shaking | None |
| Wavelength Selection | Monochromator |