missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ SPDL1 Monoclonal Antibody (K06314_12F11), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Tekniske data
| Antigen | SPDL1 |
|---|---|
| Clone | K06314_12F11 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|
30207060
|
Invitrogen™
MA556222 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30197146
|
Invitrogen™
MA555842 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
SPDL1 Monoclonal antibody specifically detects SPDL1 in Human samples. It is validated for Western BlotTekniske data
| SPDL1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1700018I02Rik; 2600001J17Rik; 2810049B11Rik; AA409762; arsenite-related gene 1 protein; Ccdc99; coiled-coil domain containing 99; coiled-coil domain-containing protein 99; coiled-coil domain-containing protein 99 {ECO:0000255; HAMAP-Rule:MF_03041}; hSpindly; protein Spindly; RGD1306908; Rhabdomyosarcoma antigen MU-RMS-40.4A; rrhabdomyosarcoma antigen protein MU-RMS-40.4A; SPDL1; Spindle apparatus coiled-coil domain-containing protein 1; spindle apparatus coiled-coil domain-containing protein 1 {ECO:0000255; spindle apparatus coiled-coil protein 1; spindly homolog | |
| Human SPDL1 peptide (proprietary range information). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SPDL1 |
| K06314_12F11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96EA4 | |
| 54908 | |
| IgG2a | |
| Protein A | |
| Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel