missing translation for 'onlineSavingsMsg'
Learn More

Schneider Gerd™ Spatula

Product Code. 10366314
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10366314

Brand: Schneider Gerd™ 3162

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use this spatula for a variety of applications. Schneider Gerd™ Spatulas are ideal for use with a wide range of compounds.

  • Laboratory spatula
  • Ideal for working with powders, stirring and mixing
  • Perfect for miniscule amounts of chemicals

Order Info

LengthMetric

Specifications

Quantity 1 Pk.
Product Type Spatula
Width (Metric) Blade 7.5 mm
Dimensions (L x W) 150 x 7.5 mm
Length (Metric) 150 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.