missing translation for 'onlineSavingsMsg'
Learn More

SPATA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen SPATA6
Concentration 0.32 mg/mL
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16859193
View Documents
Proteintech
11849-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16849193
View Documents
Proteintech
11849-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

May play a role in spermatid maturation or sperm function.
Specifications

Specifications

SPATA6
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
Q3U6K5, Q99MU5, Q9NWH7
171413, 54558, 67946
SPATA6 Fusion Protein Ag2413
Primary
-20°C
SPATA6
0.32 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
HASH, SPATA6, spermatogenesis associated 6, SRF 1, SRF1
Spata6
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.