Learn More
SPATA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11849-1-AP-20UL
 View more versions of this product
Product Code. 16859193
  • £135.00 / 20µL
Description
May play a role in spermatid maturation or sperm function.
Specifications
Specifications
SPATA6
Polyclonal
Unconjugated
SPATA6
HASH, SPATA6, spermatogenesis associated 6, SRF 1, SRF1
Rabbit
Antigen Affinity Chromatography
RUO
171413, 54558, 67946
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3U6K5, Q99MU5, Q9NWH7
Spata6
SPATA6 Fusion Protein Ag2413
20 μL
Primary
Human, Mouse, Rat