missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAST Mouse anti-Human, Unconjugated, Clone: 1F5A7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SPAST |
|---|---|
| Clone | 1F5A7 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889945
|
Proteintech
67361-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879945
|
Proteintech
67361-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SPAST antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SPAST. This antibody recognizes human antigen. The SPAST antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SPAST | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 6683 | |
| Primary |
| 1F5A7 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title