Learn More
SPAST Mouse anti-Human, Unconjugated, Clone: 1F5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67361-1-IG-20UL
This item is not returnable.
View return policy
Description
The SPAST antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SPAST. This antibody recognizes human antigen. The SPAST antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SPAST | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1F5A7 | |
| Unconjugated | |
| 20 μL | |
| 6683 | |
| IgG2b |