missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare Spacer For SE 600/SE 400 Vertical Gel Units
Manufactured using virtually unbreakable, durable plastic that is resistant to acids, basis and all common buffer systems
Brand: GE Healthcare SE6419-2-1.0
Code : 04
Additional Details : Weight : 0.80000kg
Description
All combs and spacers are moulded or fabricated of durable plastics highly resistant to acids, bases and all common buffer systems.They are lightweight and virtually unbreakable when used with reasonable care.Preparative combs have adjustable comb back, required to convert any 25mm depth comb to 10mm or 15mm depth.
Specifications
SE 600, SE 400 Vertical Gel Unit | |
2 Pack | |
1 mm |
Spacer | |
120 mm |