missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare Spacer For SE 600/SE 400 Vertical Gel Units
Manufactured using virtually unbreakable, durable plastic that is resistant to acids, basis and all common buffer systems
£60.65 - £61.65
Specifications
For Use With (Equipment) | SE 600, SE 400 Vertical Gel Unit |
---|---|
Type | Spacer |
Quantity | 2 Pack |
Length (Metric) | 120 mm |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
12064627
|
GE Healthcare
SE6419-2-1.0 |
1 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12024727
|
GE Healthcare
SE6419-2-.75 |
0.75 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
All combs and spacers are moulded or fabricated of durable plastics highly resistant to acids, bases and all common buffer systems.They are lightweight and virtually unbreakable when used with reasonable care.
Preparative combs have adjustable comb back, required to convert any 25mm depth comb to 10mm or 15mm depth.
Specifications
SE 600, SE 400 Vertical Gel Unit | |
Spacer | |
2 Pack | |
120 mm |