missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Long Handle Sampling Spoons
Description
Designed to gather small amounts of sample material, especially in hard to reach containers.
- Long handle reaches into jars, bottles and other confined areas
- Minimizes the potential of personal contact with sampled material
- FDA grade polypropylene plastic, safe for food, drug and cosmetic use
Specifications
Specifications
| Quantity | 12/Pk. |
| Description | Assortment: 3 of each size (0.25, 0.5, 1, 3 tsp.) |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1.25, 2.46, 4.93, 14.70 mL |
| Color | White |
| Certifications/Compliance | FDA |
| Disposable | Single-use |
| Product Type | Spoon |
| For Use With (Application) | Designed to gather small amounts of sample material from jars and bottles, minimize the potential of personal contact with sampled materials |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?