Learn More
Bel-Art™ SP Scienceware™ Micro-Mill™ Grinder Blades
Product Code. 11316354
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material unitSize
11316354 Stainless Steel Each
1 options
Product Code. 11316354 Supplier Bel-Art™ Supplier No. H372570000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement blades for MicroMill grinder
Stainless-steel or hard-faced blades.
Certifications
CE Marked
Specifications
Product Type Replacement Blade
For Use With (Application) Ideal for processing minerals, pharmaceuticals, pigment blends and other solid samples
For Use With (Equipment) MicroMill grinder
Material Stainless Steel
Speed 12,000 rpm