missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX22 Antibody (PCRP-SOX12-1E4), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | SOX22 |
|---|---|
| Clone | PCRP-SOX12-1E4 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry : 1-2 μg/million cells |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671154
|
Novus Biologicals
NBP3-23900-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633904
|
Novus Biologicals
NBP3-23900-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX22 Monoclonal antibody specifically detects SOX22 in Human samples. It is validated for Flow CytometrySpecifications
| SOX22 | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Protein SOX-22, SOX-22 protein, SOX22transcription factor SOX-12, SRY (sex determining region Y)-box 12, SRY (sex determining region Y)-box 22, SRY-related HMG-box gene 22 | |
| Recombinant fragment (around aa31-110) of human SOX22 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-SOX12-1E4 | |
| Flow Cytometry : 1-2 μg/million cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6666 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title