missing translation for 'onlineSavingsMsg'
Learn More

SOX22 Antibody (PCRP-SOX12-1E4), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-23900-20ug

 View more versions of this product

Product Code. 18633904

  • £178.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SOX22 Monoclonal antibody specifically detects SOX22 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SOX22
Monoclonal
0.2 mg/mL
Flow Cytometry : 1-2 μg/million cells
Protein SOX-22, SOX-22 protein, SOX22transcription factor SOX-12, SRY (sex determining region Y)-box 12, SRY (sex determining region Y)-box 22, SRY-related HMG-box gene 22
Recombinant fragment (around aa31-110) of human SOX22 protein
20 μg
Primary
Human
Purified
Flow Cytometry
PCRP-SOX12-1E4
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
6666
Store at 4°C.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.