Learn More
Invitrogen™ SOX2 Recombinant Rat Monoclonal Antibody (Btjce), Alexa Fluor™ Plus 594, Invitrogen™
Rat Recombinant Monoclonal Antibody
£188.00 - £360.00
Specifications
| Antigen | SOX2 |
|---|---|
| Clone | Btjce |
| Concentration | 1.0 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30182281
|
Invitrogen™
740013TP59420UG |
20 μL |
£188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30194970
|
Invitrogen™
740013TP594 |
50 μL |
£360.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Applications Tested: This Btjce antibody has been tested by immunohistochemistry in mouse brain tissue, immunocytochemistry and flow cytometric analysis in NTERA-2 cells. This may be used for immunocytochemistry at 2 μg/mL and for flow cytometry at 0.5 μg per test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 590 nm; Emission: 618 nm; Laser: Yellow Laser Filtration: 0.2 μm post-manufacturing filtered.
SOX2 Recombinant Monoclonal antibody specifically detects SOX2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)Specifications
| SOX2 | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8 | |
| ANOP3; cb236; Delta EF2a; lcc; MCOPS3; MCOPS3 (Microphthalmia Syndromic type 3); RGD1565646; sex determining region Y-box 2; SOX 2; SOX2; Sox-2; soxp; SRY (sex determining region Y) box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY related HMG box 2; SRY-box 2; SRY-box 2; SOX2; SRY-box containing gene 2; SRY-box transcription factor 2; SRY-related HMG-box gene 2; transcription factor SOX2; transcription factor SOX-2; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| Btjce | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Alexa Fluor Plus 594 | |
| Rat | |
| Human, Mouse | |
| P48431, P48432 | |
| 20674, 6657 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| SOX2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.