missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SOX18 Antibody (SOX18/3271), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | SOX18 |
|---|---|
| Clone | SOX18/3271 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043679
|
Novus Biologicals™
NBP3-26714-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043732
|
Novus Biologicals™
NBP3-26714-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX18 Monoclonal antibody specifically detects SOX18 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| SOX18 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
| Recombinant human SOX18 protein fragment (around aa 50-250) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| SOX18/3271 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 54345 | |
| IgG2 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title