missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SOX18 Antibody (SOX18/3271), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26714-20ug
This item is not returnable.
View return policy
Description
SOX18 Monoclonal antibody specifically detects SOX18 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SOX18 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
| Recombinant human SOX18 protein fragment (around aa 50-250) (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SOX18/3271 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 54345 | |
| Store at 4°C. | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction