Learn More
Sotalol hydrochloride, 98%, Thermo Scientific™
Potent beta adrenergic antagonist
£53.00 - £662.98
Chemical Identifiers
| CAS | 959-24-0 |
|---|---|
| Molecular Formula | C12H21ClN2O3S |
| Molecular Weight (g/mol) | 308.82 |
| MDL Number | MFCD00242937 |
| InChI Key | VIDRYROWYFWGSY-UHFFFAOYNA-N |
| Synonym | sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine |
| PubChem CID | 66245 |
| ChEBI | CHEBI:9207 |
| IUPAC Name | hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride |
| SMILES | [H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15405949
|
Thermo Scientific Alfa Aesar
J63772.MC |
100mg |
£54.20 £53.00 / 100mg Save £1.20 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15485939
|
Thermo Scientific Alfa Aesar
J63772.03 |
1g |
£340.00 £333.00 / 1g Save £7.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15495939
|
Thermo Scientific Alfa Aesar
J63772.06 |
5g |
£678.00 £662.98 / 5g Save £15.02 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsSotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.
Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 959-24-0 | |
| 308.82 | |
| VIDRYROWYFWGSY-UHFFFAOYNA-N | |
| 66245 | |
| hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride |
| C12H21ClN2O3S | |
| MFCD00242937 | |
| sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine | |
| CHEBI:9207 | |
| [H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 |
Specifications
| 959-24-0 | |
| MFCD00242937 | |
| sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine | |
| VIDRYROWYFWGSY-UHFFFAOYNA-N | |
| hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride | |
| 66245 | |
| 308.82 | |
| Sotalol hydrochloride |
| C12H21ClN2O3S | |
| 14,8728 | |
| Soluble in phosphate buffered saline,DMSO,ethanol,water,and methanol. | |
| [H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 | |
| 308.82 | |
| CHEBI:9207 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 213-496-0
missing translation for 'rtecsNumber' : PB0826000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.