missing translation for 'onlineSavingsMsg'
Learn More

Sotalol hydrochloride, 98%, Thermo Scientific™

Potent beta adrenergic antagonist

Brand:  Thermo Scientific Alfa Aesar J63772.03

Quantity 1g

 View more versions of this product

Product Code. 15485939

  • ONLINE EXCLUSIVE PRICE
    £340.00
    £333.00 / 1g
    Save £7.00
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.

Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

959-24-0
308.82
VIDRYROWYFWGSY-UHFFFAOYNA-N
66245
hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
C12H21ClN2O3S
MFCD00242937
sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
CHEBI:9207
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

Specifications

959-24-0
MFCD00242937
14,8728
Soluble in phosphate buffered saline,DMSO,ethanol,water,and methanol.
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1
308.82
CHEBI:9207
98%
C12H21ClN2O3S
1g
sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
VIDRYROWYFWGSY-UHFFFAOYNA-N
hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
66245
308.82
Sotalol hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.