missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORCS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SORCS1 |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16882155
|
Proteintech
23002-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16872155
|
Proteintech
23002-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SORCS1 is one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein.Specifications
| SORCS1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8WY21, Q9JLC4 | |
| 114815, 58178 | |
| SORCS1 Fusion Protein Ag19199 | |
| Primary | |
| -20°C | |
| SORCS1 |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hSorCS, SORCS, SORCS1 | |
| Sorcs1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title