missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORCS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23002-1-AP-20UL
This item is not returnable.
View return policy
Description
SORCS1 is one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein.Specifications
| SORCS1 | |
| Polyclonal | |
| Unconjugated | |
| SORCS1 | |
| hSorCS, SORCS, SORCS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114815, 58178 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WY21, Q9JLC4 | |
| Sorcs1 | |
| SORCS1 Fusion Protein Ag19199 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction