Learn More
Thermo Scientific™ Sorbitol MacConkey Agar with Cefixime Tellurite
Description
Efficiently isolate pathogenic Escherichia coli O157:H7 from clinical samples with Thermo Scientific™ Sorbitol MacConkey Agar with Cefixime Tellurite. Escherichia coli O157:H7 is recognised as a cause of haemorrhagic colitis, an illness characterized by bloody diarrhoea and severe abdominal pain, and haemolytic uraemic syndrome, and, as such, it is a significant human pathogen.
The efficiency of Sorbitol MacConkey Agar has been confirmed by March and Ratnam1. These workers reported a sensitivity of 100% and a specificity of 85% and recommended the medium as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 10°C |
| Description | Sorbitol MacConkey with Cefixime Tellurite |
| Format | 90mm Monoplate |
| Product Type | Agar |
| Fill Volume | 19 mL |
| Fill Weight | 19g |
| Quantity | 10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.