missing translation for 'onlineSavingsMsg'
Learn More

Sorbic acid potassium salt, ∽99%, MP Biomedicals™

Product Code. 11436161 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g

Product Code. 11436161

Brand: MP Biomedicals 0219114990

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 590-00-1
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.22
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
Synonym potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
PubChem CID 23676745
ChEBI CHEBI:77868
IUPAC Name potassium (2E,4E)-hexa-2,4-dienoate
SMILES [K+].C\C=C\C=C\C([O-])=O

Specifications

Color White
Density 1.36g/mL
Assay Percent Range 99%
Quantity 500 g
Formula Weight 150.2
Physical Form Crystalline Powder
Chemical Name or Material Sorbic acid potassium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.