accessibility menu, dialog, popup

UserName

CAS RN 590-00-1

KCH3OO(E)(E)

CAS RN 590-00-1

IUPAC Name: potassium (2E,4E)-hexa-2,4-dienoate
Synonyms: potassium sorbate
Molecular Weight (g/mol): 150.22
Molecular Formula: C6H7KO2
InChi Key: CHHHXKFHOYLYRE-STWYSWDKSA-M
SMILES: [K+].C\C=C\C=C\C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results