Learn More
Invitrogen™ Sonepcizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559796
 View more versions of this product
Product Code. 30284328
  • £1002.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sonepcizumab, also referred to as LT1009, is a humanized monoclonal antibody that targets sphingosine 1-phosphate (S1P), a signaling lipid active in promoting angiogenesis and tumor growth. Sphingosine-1-phosphate (S1P) is a critical lysolipid involved in multiple biological functions, including heart development, immune response, and hearing. It is a metabolite from cell membrane sphingolipids and is primarily regulated through its binding to specific G protein-coupled receptors named S1P receptors (S1PRs). These receptors are ubiquitously expressed in the human body, influencing various physiological and pathological processes. The signaling pathways initiated by S1P-S1PR interactions are crucial for immune cell trafficking, activation, and adaptive immune responses. Studies also highlight the role of S1P in regulating vascular stability, endothelial cell function, and as a mediator in inflammatory responses. The SPNS2 gene encodes a transporter protein essential for the active secretion of S1P, further emphasizing its relevance in maintaining physiological homeostasis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sonepcizumab Humanized
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG1