missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sonepcizumab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Sonepcizumab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284017
GREENER_CHOICE
View Documents
Invitrogen™
MA559795
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284328
View Documents
Invitrogen™
MA559796
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Sonepcizumab, also referred to as LT1009, is a humanized monoclonal antibody that targets sphingosine 1-phosphate (S1P), a signaling lipid active in promoting angiogenesis and tumor growth. Sphingosine-1-phosphate (S1P) is a critical lysolipid involved in multiple biological functions, including heart development, immune response, and hearing. It is a metabolite from cell membrane sphingolipids and is primarily regulated through its binding to specific G protein-coupled receptors named S1P receptors (S1PRs). These receptors are ubiquitously expressed in the human body, influencing various physiological and pathological processes. The signaling pathways initiated by S1P-S1PR interactions are crucial for immune cell trafficking, activation, and adaptive immune responses. Studies also highlight the role of S1P in regulating vascular stability, endothelial cell function, and as a mediator in inflammatory responses. The SPNS2 gene encodes a transporter protein essential for the active secretion of S1P, further emphasizing its relevance in maintaining physiological homeostasis.
Specifications

Specifications

Sonepcizumab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.