missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Somatostatin Recombinant Rabbit Monoclonal Antibody (021), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£74.50 - £359.00
Specifications
| Antigen | Somatostatin |
|---|---|
| Clone | 021 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019611
|
Invitrogen™
MA552272 |
20 μL |
£74.50
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019574
![]() |
Invitrogen™
MA552271 |
100 μL |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Somatostatin Recombinant Monoclonal antibody specifically detects Somatostatin in Human samples. It is validated for ELISASpecifications
| Somatostatin | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14 | |
| A synthetic peptide corresponding to the Human Somatostatin. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SST |
| 021 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P61278 | |
| 6750 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title