missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Somatostatin Recombinant Rabbit Monoclonal Antibody (021), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552272
This item is not returnable.
View return policy
Description
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.
Specifications
| Somatostatin | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with no preservative | |
| P61278 | |
| SST | |
| A synthetic peptide corresponding to the Human Somatostatin. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 021 | |
| Unconjugated | |
| SST | |
| Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction