missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R1/SSTR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £375.00
Specifications
| Antigen | Somatostatin R1/SSTR1 |
|---|---|
| Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18293460
|
Novus Biologicals
NB300-120SS |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18274160
|
Novus Biologicals
NB300-120 |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Somatostatin R1/SSTR1 Polyclonal specifically detects Somatostatin R1/SSTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Somatostatin R1/SSTR1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| Whole antisera with 0.05% Sodium Azide | |
| G-protein coupled receptor, somatostatin receptor 1, somatostatin receptor type 1, SRIF-2, SS1R, SS-1-R, SS1-R, SSR1 | |
| SSTR1 | |
| IgG | |
| Unpurified | |
| Specific to Somatostatin Receptor I (SSR1) and does not react with Somatostatin Receptor II (SSR2). |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| P28646 | |
| 6751 | |
| A synthetic peptide from the C-terminus of rat Somatostatin Receptor 1, conjugated to cysteine. [UniProt# P28646] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title