missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R1/SSTR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-120SS
This item is not returnable.
View return policy
Description
Somatostatin R1/SSTR1 Polyclonal specifically detects Somatostatin R1/SSTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Somatostatin R1/SSTR1 | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:10-1:500 | |
| P28646 | |
| SSTR1 | |
| A synthetic peptide from the C-terminus of rat Somatostatin Receptor 1, conjugated to cysteine. [UniProt# P28646] | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific to Somatostatin Receptor I (SSR1) and does not react with Somatostatin Receptor II (SSR2). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| G-protein coupled receptor, somatostatin receptor 1, somatostatin receptor type 1, SRIF-2, SS1R, SS-1-R, SS1-R, SSR1 | |
| Rabbit | |
| 60 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6751 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction