missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solid Natural Rubber Stopper

Prevents liquid chemical leaks or escape from container

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11597582

  • £26.95 / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket

Description

Description

  • Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
  • Preferable for applications that require tighter seal or greater degree of chemical resistance
  • Prevent contamination of samples by protecting contents of laboratory glassware from air
  • Allows experimenters to shake or mix solutions without spilling them
Specifications

Specifications

Red
54mm
1/Ea.
46/55 IRHD
Solid Stopper
92 mm
Natural Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers