missing translation for 'onlineSavingsMsg'
Learn More

Sodium tin(IV) oxide hydrate, Reagent Grade

Product Code. 11381549
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381549 100 g 100g
11371549 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381549 Supplier Thermo Scientific Alfa Aesar Supplier No. 014021.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12058-66-1 | Na2O3Sn | 212.69 g/mol

Sodium tin(IV) oxide is used as a mordant, fire retardant textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12058-66-1
Molecular Formula Na2O3Sn
Molecular Weight (g/mol) 212.69
MDL Number MFCD00014241
InChI Key TVQLLNFANZSCGY-UHFFFAOYSA-N
Synonym sodium stannate, sodium tin oxide, tin sodium oxide, sodium stannate iv, disodium tin trioxide, unii-jc8111zn4w, stannate sno32-, disodium, disodium stannate, disodium dioxido oxo tin, natriumstannat
PubChem CID 25501
SMILES [Na+].[Na+].[O-][Sn]([O-])=O

Specifications

Melting Point 140°C (decomposition)
Physical Form Crystalline
Quantity 100 g
Merck Index 14,8677
Solubility Information Slightly soluble in water.
IUPAC Name disodium;dioxido(oxo)tin
Formula Weight 212.67 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Sodium tin(IV) oxide hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium tin(IV) oxide hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Sodium tin(IV) oxidehydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.