CAS RN 12058-66-1
IUPAC Name:
disodium oxostannanebis(olate)
Synonyms:
sodium stannate
Molecular Weight (g/mol):
212.69
Molecular Formula:
Na2O3Sn
InChi Key:
TVQLLNFANZSCGY-UHFFFAOYSA-N
SMILES:
[Na+].[Na+].[O-][Sn]([O-])=O
1
–
1
of
1
results
1
Sodium tin(IV) oxide hydrate, Reagent Grade
CAS: 12058-66-1 Molecular Formula: Na2O3Sn Molecular Weight (g/mol): 212.69 MDL Number: MFCD00014241 InChI Key: TVQLLNFANZSCGY-UHFFFAOYSA-N Synonym: sodium stannate,sodium tin oxide,tin sodium oxide,sodium stannate iv,disodium tin trioxide,unii-jc8111zn4w,stannate sno32-, disodium,disodium stannate,disodium dioxido oxo tin,natriumstannat PubChem CID: 25501 SMILES: [Na+].[Na+].[O-][Sn]([O-])=O