Learn More
Sodium tin(IV) oxide hydrate, Reagent Grade, Thermo Scientific Chemicals
£26.90 - £73.90
Chemical Identifiers
CAS | 12058-66-1 |
---|---|
Molecular Formula | Na2O3Sn |
Molecular Weight (g/mol) | 212.69 |
MDL Number | MFCD00014241 |
InChI Key | TVQLLNFANZSCGY-UHFFFAOYSA-N |
Synonym | sodium stannate, sodium tin oxide, tin sodium oxide, sodium stannate iv, disodium tin trioxide, unii-jc8111zn4w, stannate sno32-, disodium, disodium stannate, disodium dioxido oxo tin, natriumstannat |
PubChem CID | 25501 |
SMILES | [Na+].[Na+].[O-][Sn]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381549
|
Thermo Scientific Alfa Aesar
014021.22 |
100 g |
£26.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371549
|
Thermo Scientific Alfa Aesar
014021.36 |
500 g |
£73.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium tin(IV) oxide is used as a mordant, fire retardant textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12058-66-1 | |
212.69 | |
TVQLLNFANZSCGY-UHFFFAOYSA-N | |
25501 |
Na2O3Sn | |
MFCD00014241 | |
sodium stannate, sodium tin oxide, tin sodium oxide, sodium stannate iv, disodium tin trioxide, unii-jc8111zn4w, stannate sno32-, disodium, disodium stannate, disodium dioxido oxo tin, natriumstannat | |
[Na+].[Na+].[O-][Sn]([O-])=O |
Specifications
140°C (decomposition) | |
Crystalline | |
MFCD00014241 | |
sodium stannate, sodium tin oxide, tin sodium oxide, sodium stannate iv, disodium tin trioxide, unii-jc8111zn4w, stannate sno32-, disodium, disodium stannate, disodium dioxido oxo tin, natriumstannat | |
TVQLLNFANZSCGY-UHFFFAOYSA-N | |
disodium;dioxido(oxo)tin | |
25501 | |
Reagent | |
Air Sensitive |
12058-66-1 | |
Na2O3Sn | |
14,8677 | |
Slightly soluble in water. | |
[Na+].[Na+].[O-][Sn]([O-])=O | |
212.69 | |
212.67 (Anhydrous) | |
Odorless | |
Sodium tin(IV) oxide hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 235-030-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only