missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloroaluminate

Product Code. 11398027
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398027 25 g 25g
11308037 5 g 5g
11318037 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398027 Supplier Thermo Scientific Alfa Aesar Supplier No. 022375.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7784-16-9 | AlCl4Na | 191.771 g/mol

Sodium tetrachloroaluminate is used as Friedel-Crafts acylating agent , dehydrating and dehydrogenating agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-16-9
Molecular Formula AlCl4Na
Molecular Weight (g/mol) 191.771
MDL Number MFCD00050361
InChI Key CMLRNXGMTZKKSR-UHFFFAOYSA-J
Synonym sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis
PubChem CID 16699350
IUPAC Name sodium;tetrachloroalumanuide
SMILES [Na+].[Al-](Cl)(Cl)(Cl)Cl

Specifications

Melting Point 185°C
Color Yellow
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in water
Formula Weight 191.78
Sensitivity Moisture sensitive
Appearance Pale yellow powder
Density 2.01 g/cm3
Chemical Name or Material Sodium tetrachloroaluminate
compliance-icons
Product Identifier
  • Sodium tetrachloroaluminate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Aluminium sodium tetrachloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.