accessibility menu, dialog, popup

UserName

CAS RN 7784-16-9

NaAlClClClCl

CAS RN 7784-16-9

IUPAC Name: sodium tetrachloroalumanuide
Synonyms: sodium tetrachloroaluminate(1-)
Molecular Weight (g/mol): 191.77
Molecular Formula: AlCl4Na
InChi Key: CMLRNXGMTZKKSR-UHFFFAOYSA-J
SMILES: [Na+].Cl[Al-](Cl)(Cl)Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results