Learn More
Sodium tetrachloroaluminate, Thermo Scientific Chemicals
£30.03 - £917.00
Chemical Identifiers
CAS | 7784-16-9 |
---|---|
Molecular Formula | AlCl4Na |
Molecular Weight (g/mol) | 191.771 |
MDL Number | MFCD00050361 |
InChI Key | CMLRNXGMTZKKSR-UHFFFAOYSA-J |
Synonym | sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis |
PubChem CID | 16699350 |
IUPAC Name | sodium;tetrachloroalumanuide |
SMILES | [Na+].[Al-](Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308037
|
Thermo Scientific Alfa Aesar
022375.06 |
5 g | |||||||
11398027
|
Thermo Scientific Alfa Aesar
022375.14 |
25 g |
£243.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318037
|
Thermo Scientific Alfa Aesar
022375.22 |
100 g |
£917.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium tetrachloroaluminate is used as Friedel-Crafts acylating agent , dehydrating and dehydrogenating agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-16-9 | |
191.771 | |
CMLRNXGMTZKKSR-UHFFFAOYSA-J | |
16699350 | |
[Na+].[Al-](Cl)(Cl)(Cl)Cl |
AlCl4Na | |
MFCD00050361 | |
sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis | |
sodium;tetrachloroalumanuide |
Specifications
7784-16-9 | |
Powder | |
MFCD00050361 | |
CMLRNXGMTZKKSR-UHFFFAOYSA-J | |
sodium;tetrachloroalumanuide | |
16699350 | |
Pale yellow powder |
Yellow | |
AlCl4Na | |
sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis | |
[Na+].[Al-](Cl)(Cl)(Cl)Cl | |
191.771 | |
191.78 | |
Sodium tetrachloroaluminate |
Safety and Handling
H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
missing translation for 'dotInformation' : Hazard Class: 8, Packing Group: III
missing translation for 'einecsNumber' : 232-050-6
RUO – Research Use Only