missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Alpha 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94669-0.1ml
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Alpha 2 Polyclonal antibody specifically detects Sodium Potassium ATPase Alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Sodium Potassium ATPase Alpha 2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500 | |
| alpha-A(+) catalytic polypeptide, ATPase, Na+/K+ transporting, alpha 2 (+) polypeptide, ATPase, Na+/K+ transporting, alpha 2 polypeptide, EC 3.6.3, EC 3.6.3.9, KIAA0778, MGC59864, MHP2, migraine, hemiplegic 2, Na(+)/K(+) ATPase alpha-2 subunit, Na+/K+ ATPase, alpha-B polypeptide, Sodium pump subunit alpha-2, sodium/potassium-transporting ATPase alpha-2 chain, sodium/potassium-transporting ATPase subunit alpha-2, sodium-potassium ATPase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human ATP1A2 (NP_000693.1). MGRGAGREYSPAATTAENGGGKKKQKEKELDELKKEVAMDDHKLSLDELGRKYQVDLSKGLTNQRAQDVL | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 477 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction