missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen DL-malate, 98%

Product Code. 15473187
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473187 50 g 50g
15483187 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473187 Supplier Thermo Scientific Alfa Aesar Supplier No. A19428.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57467-17-1 | C4H5NaO5 | 156.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57467-17-1
Molecular Formula C4H5NaO5
Molecular Weight (g/mol) 156.07
MDL Number MFCD00798569
InChI Key DOJOZCIMYABYPO-UHFFFAOYNA-M
Synonym sodium hydrogen malate, monosodium malate, sodium malate, l-malic acid, --sodium hydrogen malate, l--malic acid disodium salt, sodium 3-carboxy-3-hydroxypropanoate, 2-hydroxysuccinic acid 1-sodium salt, sodium 3,4-dihydroxy-4-oxo-butanoate, 2-hydroxybutanedioic acid 4-sodium salt
PubChem CID 23678821
IUPAC Name sodium;3,4-dihydroxy-4-oxobutanoate
SMILES [Na+].OC(CC([O-])=O)C(O)=O

Specifications

Melting Point 210°C (decomposition)
Quantity 50 g
Formula Weight 156.07
Percent Purity 98%
Chemical Name or Material Sodium hydrogen DL-malate
compliance-icons
Product Identifier
  • Sodium hydrogen DL-malate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydrogen DL-malate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.