accessibility menu, dialog, popup

UserName

CAS RN 57467-17-1

OOHOHOONa

CAS RN 57467-17-1

IUPAC Name: sodium 3-carboxy-3-hydroxypropanoate
Synonyms: sodium malate
Molecular Weight (g/mol): 156.07
Molecular Formula: C4H5NaO5
InChi Key: DOJOZCIMYABYPO-UHFFFAOYNA-M
SMILES: [Na+].OC(CC([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results