missing translation for 'onlineSavingsMsg'
Learn More

Sodium galacturonate anhydrous, 98%

Product Code. 15418189
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418189 25 g 25g
15408189 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418189 Supplier Thermo Scientific Alfa Aesar Supplier No. J66724.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14984-39-5 | C6H9NaO7 | 216.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14984-39-5
Molecular Formula C6H9NaO7
Molecular Weight (g/mol) 216.121
MDL Number MFCD06201013
InChI Key MSXHSNHNTORCAW-NSBQNPTMSA-M
Synonym d-+-galacturonic acid sodium salt
PubChem CID 122174033
IUPAC Name sodium;(4S)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
SMILES C1(C(C(OC(C1O)O)C(=O)[O-])O)O.[Na+]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Formula Weight 216.12
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium galacturonate, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.