accessibility menu, dialog, popup

UserName

CAS RN 14984-39-5

OHOHOOOOHOHNa

CAS RN 14984-39-5

IUPAC Name: sodium 3,4,5,6-tetrahydroxyoxane-2-carboxylate
Synonyms: sodium 3,4,5,6-tetrahydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 216.12
Molecular Formula: C6H9NaO7
InChi Key: MSXHSNHNTORCAW-UHFFFAOYNA-M
SMILES: [Na+].OC1OC(C(O)C(O)C1O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results