missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium dodecamolybdophosphate hydrate, 90+%

Product Code. 11404593
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404593 500 g 500g
11494583 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404593 Supplier Alfa Aesar™ Supplier No. A11551.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1313-30-0
Molecular Formula Mo12Na3O40P
Molecular Weight (g/mol) 1891.18
MDL Number MFCD00036307
InChI Key FROPCQVXXZDEBU-UHFFFAOYSA-M
IUPAC Name Sodium dodecamolybdophosphate hydrate
SMILES *

Specifications

Quantity 500 g
Merck Index 14,8664
Formula Weight 1891.22 (Anhydrous)
Percent Purity ≥90%
Chemical Name or Material Sodium dodecamolybdophosphate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.