accessibility menu, dialog, popup

UserName

CAS RN 1313-30-0

A(a)

CAS RN 1313-30-0

IUPAC Name: molybdenum hydroxide oxidandiide phosphanetriide
Synonyms: phosphomolybdic acid
Molecular Weight (g/mol): 159.93
Molecular Formula: HMoO2P
InChi Key: FROPCQVXXZDEBU-UHFFFAOYSA-M
SMILES: [OH-].[O--].[P-3].[Mo]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results