missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Hydrate, ≥99.0% (Calc. based on dry substance, T), Honeywell™

Product Code. p-7120775 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 15618460

Brand: Honeywell Research Chemicals 713611KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (calc. based on dry substance, T), 9-10 mol/mol water

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24551-51-7
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.138
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

pH 12 (at 25°C)
Physical Form Crystalline
Quantity 1 kg
Linear Formula Na2CO3 · aq
UN Number NONH for all modes of transport
Solubility Information 142.2g/L (at 20°C)
Formula Weight 105.99g/mol (anhydrous basis)
Percent Purity ≥99.0% (calc. based on dry substance, T)
Packaging Poly bottle
Chemical Name or Material Sodium carbonate hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.