accessibility menu, dialog, popup

UserName

OOOH2OH2OH2OH2OH2OH2OH2OH2OH2OH2ONaNa

CAS RN 24551-51-7

IUPAC Name: disodium decahydrate carbonate
Synonyms: disodium decahydrate carbonate
Molecular Weight (g/mol): 286.14
Molecular Formula: CH20Na2O13
InChi Key: XYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)