missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Hydrate, ≥99.0% (Calc. based on dry substance, T), Honeywell™

Product Code. 15628460 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15628460 5 kg Poly bottle 5kg
15618460 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15628460 Supplier Honeywell Research Chemicals Supplier No. 713615KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (calc. based on dry substance, T), 9-10 mol/mol water

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24551-51-7
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.138
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

pH 12 (at 25°C)
Physical Form Crystalline
Quantity 5 kg
Linear Formula Na2CO3 · aq
UN Number NONH for all modes of transport
Solubility Information 142.2g/L (at 20°C)
Formula Weight 105.99g/mol (anhydrous basis)
Percent Purity ≥99.0% (calc. based on dry substance, T)
Packaging Poly bottle
Chemical Name or Material Sodium carbonate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.