missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube

Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.

Brand:  Fisherbrand™

Code : 33

Additional Details : Weight : 0.89100kg

 View more versions of this product

Product Code. 11952188

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Features: rimmed light walls
  • Withstands: moderate warming
  • Complies to: ISO 4142
Specifications

Specifications

16 mm
125 mm
100 Pack
ISO 4142
Soda Lime Glass
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers