missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SOD (Superoxide Dismutase) Activity Assay Kit (Colorimetric)
Shop All Bio Techne ProductsSpecifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 96 Tests |
| Research Category | Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors |
| Species | Non-species specific |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?