missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-7/Nck/NAP4 Antibody (23E4), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | SOCS-7/Nck/NAP4 |
|---|---|
| Clone | 23E4 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30031294
|
Novus Biologicals
NBP3-26310-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029338
|
Novus Biologicals
NBP3-26310-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOCS-7/Nck/NAP4 Monoclonal antibody specifically detects SOCS-7/Nck/NAP4 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| SOCS-7/Nck/NAP4 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| NAP-4, NAP4SOCS-7, Nck, Ash and phospholipase C binding protein, Nck, Ash and phospholipase C gamma-binding protein, NCKAP4, Nck-associated protein 4, SOCS6, suppressor of cytokine signaling 7 | |
| A synthesized peptide derived from Human SOCS-7/Nck/NAP4 [UniProt O14512] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 23E4 | |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 30837 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title