missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-7/Nck/NAP4 Antibody (23E4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26310-100ul
This item is not returnable.
View return policy
Description
SOCS-7/Nck/NAP4 Monoclonal antibody specifically detects SOCS-7/Nck/NAP4 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| SOCS-7/Nck/NAP4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 23E4 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| NAP-4, NAP4SOCS-7, Nck, Ash and phospholipase C binding protein, Nck, Ash and phospholipase C gamma-binding protein, NCKAP4, Nck-associated protein 4, SOCS6, suppressor of cytokine signaling 7 | |
| A synthesized peptide derived from Human SOCS-7/Nck/NAP4 [UniProt O14512] | |
| 100 μL | |
| Signal Transduction | |
| 30837 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction